siRNA Details
Gene Symbol: Usp34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271196.1 | NP_001258125.1 | 68 | 8677 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_001190401, BQ179166, AK129165.
Gene Ontology Categories:
