siRNA Details
Gene Symbol: Parp4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008770809.1 | XP_008769031.1 | 21 | 2685 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Parp4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008770809.1 | XP_008769031.1 | 21 | 2685 |