siRNA Details
Gene Symbol: Rcbtb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108380.1 | NP_001101850.1 | 5 | 855 | ||
XM_008770759.1 | XP_008768981.1 | 6 | 681 | |||
GenBank mRNA | AY724518.1 | - | 4 | 501 |
