siRNA Details
Gene Symbol: Cdadc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012156.1 | NP_001012156.1 | 5 | 759 | ||
XM_006252123.1 | XP_006252185.1 | 5 | 807 | |||
XM_006252125.2 | XP_006252187.1 | 5 | 807 | |||
XM_006252126.2 | XP_006252188.1 | 4 | 710 | |||
XM_008770760.1 | XP_008768982.1 | 4 | 734 | |||
GenBank mRNA | BC083736.1 | - | 5 | 759 | 7191631 | |
FQ213068.1 | - | 5 | 778 |
