siRNA Details
Gene Symbol: Ebpl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108381.1 | NP_001101851.1 | 4 | 518 | ||
GenBank mRNA | FQ213140.1 | - | 4 | 524 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Ebpl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108381.1 | NP_001101851.1 | 4 | 518 | ||
GenBank mRNA | FQ213140.1 | - | 4 | 524 |