siRNA Details
Gene Symbol: Lmo7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001515.1 | NP_001001515.1 | 13, 14 | 2218 | ||
XM_006252397.2 | XP_006252459.1 | 13, 14 | 2454 | |||
XM_006252399.2 | XP_006252461.1 | 11, 12 | 1448 | |||
XM_006252400.2 | XP_006252462.1 | 10, 11 | 1418 | |||
XM_006252401.2 | XP_006252463.1 | 11, 12 | 1448 | |||
XM_008770874.1 | XP_008769096.1 | 15, 16 | 2551 | |||
XM_008770875.1 | XP_008769097.1 | 14, 15 | 2656 | |||
XM_008770876.1 | XP_008769098.1 | 14, 15 | 2656 | |||
XM_008770877.1 | XP_008769099.1 | 14, 15 | 2655 | |||
XM_008770878.1 | XP_008769100.1 | 12, 13 | 3117 | |||
XM_008770879.1 | XP_008769101.1 | 14, 15 | 2655 | |||
XM_008770880.1 | XP_008769102.1 | 12, 13 | 2344 | |||
XM_008770881.1 | XP_008769103.1 | 11, 12 | 2268 | |||
GenBank mRNA | AY533473.1 | - | 13, 14 | 2218 | ||
AY533474.1 | - | 11, 12 | 1216 |
