siRNA Details
Gene Symbol: Lmo7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001515.1 | NP_001001515.1 | 16 | 3284 | ||
XM_006252397.2 | XP_006252459.1 | 16 | 3520 | |||
XM_006252399.2 | XP_006252461.1 | 14 | 2514 | |||
XM_006252400.2 | XP_006252462.1 | 13 | 2484 | |||
XM_006252401.2 | XP_006252463.1 | 14 | 2514 | |||
XM_008770874.1 | XP_008769096.1 | 18 | 3617 | |||
XM_008770875.1 | XP_008769097.1 | 17 | 3722 | |||
XM_008770876.1 | XP_008769098.1 | 17 | 3722 | |||
XM_008770877.1 | XP_008769099.1 | 17 | 3721 | |||
XM_008770878.1 | XP_008769100.1 | 15 | 4183 | |||
XM_008770879.1 | XP_008769101.1 | 17 | 3721 | |||
XM_008770880.1 | XP_008769102.1 | 15 | 3410 | |||
XM_008770881.1 | XP_008769103.1 | 14 | 3334 | |||
GenBank mRNA | AY533473.1 | - | 16 | 3284 | ||
AY533474.1 | - | 14 | 2282 | |||
AY609384.1 | - | 11 | 2297 |
