siRNA Details
Gene Symbol: Adam32 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170582.1 | NP_001164053.1 | 5, 6 | 365 | ||
XM_008771361.1 | XP_008769583.1 | 5, 6 | 475 | |||
XM_008771362.1 | XP_008769584.1 | 5, 6 | 780 | |||
GenBank mRNA | CV102424.1 | - | 5, 6 | 341 |
