siRNA Details
Gene Symbol: Cars2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177684.1 | NP_001171155.1 | 5 | 480 | ||
XM_006253460.2 | XP_006253522.1 | 5 | 928 | |||
XM_006253461.2 | XP_006253523.1 | 5 | 926 | |||
XM_006253462.2 | XP_006253524.1 | 5 | 919 | |||
GenBank mRNA | BC166536.1 | - | 5 | 480 | 7191115 |
