siRNA Details
Gene Symbol: Usp19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001516.2 | NP_001001516.1 | 20 | 2829 | ||
XM_006243793.2 | XP_006243855.1 | 21 | 3147 | |||
XM_006243794.2 | XP_006243856.1 | 21 | 3144 | |||
XM_006243795.2 | XP_006243857.1 | 21 | 3141 | |||
XM_006243796.2 | XP_006243858.1 | 21 | 3123 | |||
XM_006243797.2 | XP_006243859.1 | 21 | 3146 | |||
XM_006243798.2 | XP_006243860.1 | 21 | 3143 | |||
XM_006243799.2 | XP_006243861.1 | 21 | 3140 | |||
GenBank mRNA | AY605065.1 | - | 20 | 2829 | ||
BC168243.1 | - | 20 | 3155 |
