siRNA Details
Gene Symbol: Fgd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108409.1 | NP_001101879.1 | 13 | 1688 | ||
XM_008771543.1 | XP_008769765.1 | 15 | 2410 | |||
XM_008771544.1 | XP_008769766.1 | 15 | 3018 | |||
XM_008771545.1 | XP_008769767.1 | 15 | 2112 | |||
XM_008771546.1 | XP_008769768.1 | 15 | 2081 |
