siRNA Details
Gene Symbol: Serpinb9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007732.1 | NP_001007733.1 | 5 | 681 | ||
XM_006253872.2 | XP_006253934.1 | 5 | 650 | |||
GenBank mRNA | BC079063.1 | - | 5 | 681 | 7121006 | |
FQ226618.1 | - | 2 | 216 |
