siRNA Details
Gene Symbol: Dusp22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108412.1 | NP_001101882.1 | 3, 4 | 259 | ||
XM_006253888.2 | XP_006253950.1 | 3, 4 | 207 | |||
XM_006253889.2 | XP_006253951.1 | 3, 4 | 480 |
