siRNA Details
Gene Symbol: Svil | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108416.2 | NP_001101886.2 | 38 | 7185 | ||
XM_006254030.2 | XP_006254092.1 | 42 | 7317 | |||
XM_006254031.2 | XP_006254093.1 | 40 | 9055 | |||
XM_008771737.1 | XP_008769959.1 | 42 | 9407 | |||
XM_008771738.1 | XP_008769960.1 | 42 | 6977 | |||
XM_008771739.1 | XP_008769961.1 | 41 | 7208 | |||
XM_008771740.1 | XP_008769962.1 | 41 | 6988 | |||
XM_008771741.1 | XP_008769963.1 | 42 | 7314 | |||
XM_008771742.1 | XP_008769964.1 | 41 | 7239 | |||
XM_008771743.1 | XP_008769965.1 | 41 | 7220 | |||
XM_008771744.1 | XP_008769966.1 | 40 | 7190 | |||
XM_008771745.1 | XP_008769967.1 | 40 | 7187 | |||
XM_008771746.1 | XP_008769968.1 | 41 | 7151 | |||
XM_008771747.1 | XP_008769969.1 | 40 | 7107 | |||
XM_008771748.1 | XP_008769970.1 | 40 | 6295 | |||
XM_008771749.1 | XP_008769971.1 | 39 | 6211 | |||
XM_008771750.1 | XP_008769972.1 | 36 | 5988 |
