siRNA Details
Gene Symbol: Tom1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008365.1 | NP_001008366.1 | 5, 6 | 535 | ||
XM_006255145.2 | XP_006255207.1 | 5, 6 | 627 | |||
GenBank mRNA | BC083873.1 | - | 5, 6 | 535 | 7128979 | |
FQ213905.1 | - | 2, 3 | 207 |
