siRNA Details
Gene Symbol: Terf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108448.1 | NP_001101918.1 | 10 | 1408 | ||
XM_006255564.2 | XP_006255626.1 | 12 | 1823 | |||
GenBank mRNA | FQ212165.1 | - | 9 | 1047 |
