siRNA Details
Gene Symbol: Ppil4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108457.1 | NP_001101927.1 | 4 | 323 | ||
XM_006227621.2 | XP_006227683.1 | 4 | 357 | |||
GenBank mRNA | FQ225576.1 | - | 4 | 362 |
