siRNA Details
Gene Symbol: Pnldc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025724.1 | NP_001020895.1 | 2, 3 | 167 | ||
XM_006227880.2 | XP_006227942.1 | 2, 3 | 1300 | |||
XM_008758734.1 | XP_008756956.1 | 2, 3 | 1300 | |||
GenBank mRNA | BC099137.1 | - | 2, 3 | 167 | 7456056 |
