siRNA Details
Gene Symbol: LOC100910046 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006228601.1 | XP_006228663.1 | 5 | 2063 | ||
XM_006228602.2 | XP_006228664.1 | 5 | 2060 | |||
XM_008759207.1 | XP_008757429.1 | 5 | 2578 | |||
XM_008759208.1 | XP_008757430.1 | 5 | 2042 |
