siRNA Details
Gene Symbol: Rbm42 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014159.2 | NP_001014181.2 | 6 | 704 | ||
XM_006228781.2 | XP_006228843.1 | 5 | 642 | |||
GenBank mRNA | BC079321.1 | - | 6 | 662 | 7135399 | |
BP504208.1 | - | 5 | 624 | |||
FQ222852.1 | - | 5 | 587 |
