siRNA Details
Gene Symbol: Aen | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108487.2 | NP_001101957.1 | 3 | 1046 | ||
XM_006229399.2 | XP_006229461.1 | 3 | 837 | |||
GenBank mRNA | BC166435.1 | - | 3 | 1046 | 7324389 |
