siRNA Details
Gene Symbol: Tm6sf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108490.1 | NP_001101960.1 | 10 | 970 | ||
XM_006229441.2 | XP_006229503.1 | 9 | 852 | |||
XM_008759543.1 | XP_008757765.1 | 8 | 1232 | |||
XM_008759544.1 | XP_008757766.1 | 9 | 824 |
