siRNA Details
Gene Symbol: Arap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006229897.2 | XP_006229959.1 | 31 | 4268 | ||
XM_006229898.2 | XP_006229960.1 | 31 | 5012 | |||
XM_006229899.2 | XP_006229961.1 | 31 | 5010 | |||
XM_006229900.1 | XP_006229962.1 | 29 | 3887 | |||
XM_006229901.1 | XP_006229963.1 | 28 | 3708 | |||
XM_008759810.1 | XP_008758032.1 | 32 | 5435 | |||
XM_008759811.1 | XP_008758033.1 | 30 | 4977 | |||
GenBank mRNA | BC088328.1 | - | 8 | 819 |
