siRNA Details
Gene Symbol: Acsm5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014162.4 | NP_001014184.3 | 4 | 838 | ||
XM_006230123.1 | XP_006230185.1 | 5 | 781 | |||
XM_006230126.1 | XP_006230188.1 | 5 | 781 | |||
GenBank mRNA | AY325244.1 | - | 1074 | |||
BC078915.1 | - | 4 | 837 | 7096091 |
