siRNA Details
Gene Symbol: Plekha1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001079894.1 | NP_001073363.1 | 13 | 1277 | ||
XM_006230409.2 | XP_006230471.1 | 12 | 1207 | |||
XM_006230412.2 | XP_006230474.1 | 13 | 1279 | |||
XM_006230413.2 | XP_006230475.1 | 13 | 1248 | |||
XM_008759907.1 | XP_008758129.1 | 12 | 1282 | |||
XM_008759908.1 | XP_008758130.1 | 12 | 1603 | |||
XM_008759909.1 | XP_008758131.1 | 12 | 1176 | |||
XM_008759910.1 | XP_008758132.1 | 9 | 846 | |||
GenBank mRNA | BC128752.1 | - | 13 | 1277 | 7100072 |
