siRNA Details
Gene Symbol: Osbpl5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001015024.2 | NP_001015024.2 | 19 | 2468 | ||
XM_006230828.2 | XP_006230890.1 | 19 | 2276 | |||
XM_006230829.2 | XP_006230891.1 | 19 | 2276 | |||
XM_006230830.2 | XP_006230892.1 | 19 | 2362 | |||
GenBank mRNA | BC091326.1 | - | 2 | 390 |
