siRNA Details
Gene Symbol: Osbpl5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001015024.2 | NP_001015024.2 | 8, 9 | 1167 | ||
XM_006230828.2 | XP_006230890.1 | 8, 9 | 975 | |||
XM_006230829.2 | XP_006230891.1 | 8, 9 | 975 | |||
XM_006230830.2 | XP_006230892.1 | 8, 9 | 1061 |
