siRNA Details
Gene Symbol: Kdm2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108515.1 | NP_001101985.1 | 12 | 1756 | ||
XM_006230843.2 | XP_006230905.1 | 12 | 1528 | |||
XM_006230844.2 | XP_006230906.1 | 12 | 2127 | |||
XM_008760152.1 | XP_008758374.1 | 11 | 1776 |
