siRNA Details
Gene Symbol: Sipa1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004089.1 | NP_001004089.1 | 7 | 1893 | ||
XM_006230849.2 | XP_006230911.1 | 8 | 2510 | |||
GenBank mRNA | AB091474.1 | - | 7 | 1893 | ||
FQ223350.1 | - | 8 | 2100 | |||
FQ228273.1 | - | 8 | 2079 |
