siRNA Details
Gene Symbol: Tjp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053773.1 | NP_446225.1 | 24 | 3754 | ||
XM_006231175.2 | XP_006231237.1 | 24 | 4842 | |||
XM_006231176.2 | XP_006231238.1 | 23 | 4732 | |||
XM_006231177.2 | XP_006231239.1 | 23 | 4073 | |||
XM_006231178.2 | XP_006231240.1 | 23 | 7002 | |||
XM_006231179.2 | XP_006231241.1 | 23 | 3906 | |||
XM_006231181.2 | XP_006231243.1 | 24 | 3861 | |||
XM_006231182.2 | XP_006231244.1 | 22 | 3962 | |||
XM_008760302.1 | XP_008758524.1 | 24 | 3602 | |||
GenBank mRNA | BC103479.1 | - | 24 | 3754 | 7936785 | |
U75916.1 | - | 17 | 2393 |
