siRNA Details
Gene Symbol: Smarca2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004446.1 | NP_001004446.1 | 28 | 4231 | ||
XM_006231227.2 | XP_006231289.1 | 28 | 4249 | |||
XM_006231229.2 | XP_006231291.1 | 3 | 350 | |||
XM_008760331.1 | XP_008758553.1 | 28 | 4509 | |||
XM_008760332.1 | XP_008758554.1 | 29 | 4557 | |||
XM_008760333.1 | XP_008758555.1 | 29 | 4297 | |||
XM_008760334.1 | XP_008758556.1 | 28 | 4329 | |||
XM_008760335.1 | XP_008758557.1 | 28 | 4265 | |||
XM_008760336.1 | XP_008758558.1 | 28 | 4577 | |||
XM_008760337.1 | XP_008758559.1 | 27 | 4127 | |||
XM_008760338.1 | XP_008758560.1 | 26 | 3763 | |||
XM_008760339.1 | XP_008758561.1 | 3 | 252 | |||
GenBank mRNA | AY643746.1 | - | 28 | 4231 |
