siRNA Details
Gene Symbol: Smarca2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004446.1 | NP_001004446.1 | 15, 16 | 2525 | ||
XM_006231227.2 | XP_006231289.1 | 15, 16 | 2543 | |||
XM_008760331.1 | XP_008758553.1 | 15, 16 | 2803 | |||
XM_008760332.1 | XP_008758554.1 | 16, 17 | 2851 | |||
XM_008760333.1 | XP_008758555.1 | 16, 17 | 2591 | |||
XM_008760334.1 | XP_008758556.1 | 15, 16 | 2623 | |||
XM_008760335.1 | XP_008758557.1 | 15, 16 | 2559 | |||
XM_008760336.1 | XP_008758558.1 | 15, 16 | 2871 | |||
XM_008760337.1 | XP_008758559.1 | 14, 15 | 2421 | |||
XM_008760338.1 | XP_008758560.1 | 13, 14 | 2057 | |||
GenBank mRNA | AY643746.1 | - | 15, 16 | 2525 |
