siRNA Details
Gene Symbol: Pde6c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108522.1 | NP_001101992.1 | 12 | 1512 | ||
XM_006231328.2 | XP_006231390.1 | 12 | 1747 | |||
XM_006231329.2 | XP_006231391.1 | 12 | 1747 |
