siRNA Details
Gene Symbol: Aldh18a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108524.1 | NP_001101994.1 | 13 | 1602 | ||
XM_003749108.2 | XP_003749156.1 | 12 | 1631 | |||
XM_006231344.1 | XP_006231406.1 | 12 | 1636 | |||
XM_006231345.1 | XP_006231407.1 | 12 | 1636 | |||
XM_006231346.1 | XP_006231408.1 | 12 | 1631 |
