siRNA Details
Gene Symbol: Btrc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007148.2 | NP_001007149.2 | 6 | 511 | ||
XM_006231529.2 | XP_006231591.1 | 6 | 1094 | |||
XM_006231530.2 | XP_006231592.1 | 5 | 505 | |||
XM_006231531.2 | XP_006231593.1 | 5 | 471 | |||
XM_006231535.2 | XP_006231597.1 | 5 | 356 | |||
XM_006231536.2 | XP_006231598.1 | 5 | 438 | |||
XM_006231537.2 | XP_006231599.1 | 5 | 985 | |||
XM_006231538.2 | XP_006231600.1 | 4 | 395 | |||
XM_006231539.2 | XP_006231601.1 | 4 | 1141 | |||
XM_006231541.2 | XP_006231603.1 | 4 | 855 | |||
XM_006231543.2 | XP_006231605.1 | 3 | 1063 | |||
GenBank mRNA | BC085125.1 | - | 5 | 332 | 7131883 | |
CB584165.1 | - | 6 | 524 | |||
EV771066.1 | - | 6 | 451 |
