siRNA Details
Gene Symbol: Ehmt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_212463.1 | NP_997628.1 | 18 | 2553 | ||
XM_006255954.2 | XP_006256016.2 | 18 | 2588 | |||
XM_006255955.2 | XP_006256017.2 | 17 | 2486 | |||
XM_006255956.2 | XP_006256018.2 | 17 | 2455 | |||
XM_006255957.1 | XP_006256019.1 | 19 | 2412 | |||
XM_006255958.1 | XP_006256020.1 | 19 | 2382 | |||
XM_006255959.1 | XP_006256021.1 | 18 | 2280 |
