siRNA Details
Gene Symbol: Specc1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039455.1 | NP_001034544.1 | 15, 16 | 3461 | ||
XM_006256339.2 | XP_006256401.1 | 15, 16 | 3443 | |||
GenBank mRNA | AY884296.1 | - | 15, 16 | 3461 |
