siRNA Details
Gene Symbol: Traf3ip2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001044248.1 | NP_001037713.1 | 2 | 157 | ||
XM_006256532.2 | XP_006256594.1 | 2 | 374 | |||
GenBank mRNA | BC100099.1 | - | 2 | 157 | 7376204 |
