siRNA Details
Gene Symbol: Hace1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108539.3 | NP_001102009.2 | 9, 10 | 816 | ||
XM_006256595.2 | XP_006256657.1 | 9, 10 | 1069 | |||
XM_006256597.2 | XP_006256659.1 | 9, 10 | 1069 | |||
XM_006256598.1 | XP_006256660.1 | 4, 5 | 407 |
