siRNA Details
Gene Symbol: Atg10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109505.1 | NP_001102975.1 | 2 | 82 | ||
XM_006231756.2 | XP_006231818.1 | 3 | 349 | |||
XM_006231757.2 | XP_006231819.1 | 2 | 1120 | |||
XM_006231758.2 | XP_006231820.1 | 2 | 98 | |||
XM_006231759.2 | XP_006231821.1 | 2 | 118 |
