siRNA Details
Gene Symbol: Mccc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012177.1 | NP_001012177.1 | 13, 14 | 1356 | ||
XM_008760678.1 | XP_008758900.1 | 12, 13 | 1428 | |||
GenBank mRNA | BC083581.1 | - | 13, 14 | 1356 | 7129446 |
