siRNA Details
Gene Symbol: Ankrd55 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270039.1 | NP_001256968.1 | 8 | 2249 | ||
XM_006231940.2 | XP_006232002.1 | 9 | 1981 | |||
XM_008760737.1 | XP_008758959.1 | 10 | 2018 | |||
GenBank mRNA | JQ013731.1 | - | 8 | 2249 |
