siRNA Details
Gene Symbol: Mynn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012178.1 | NP_001012178.1 | 5 | 1202 | ||
XM_006232232.2 | XP_006232294.1 | 5 | 2246 | |||
XM_006232233.2 | XP_006232295.1 | 5 | 2247 | |||
XM_006232234.2 | XP_006232296.1 | 5 | 2116 | |||
GenBank mRNA | BC089853.1 | - | 5 | 1202 | 7379696 |
