siRNA Details
Gene Symbol: Actl6a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039033.1 | NP_001034122.1 | 3 | 236 | ||
XM_006232253.2 | XP_006232315.1 | 3 | 264 | |||
XM_008760930.1 | XP_008759152.1 | 3 | 304 | |||
XM_008760931.1 | XP_008759153.1 | 3 | 280 | |||
GenBank mRNA | BC098698.1 | - | 3 | 236 | 7386860 |
