siRNA Details
Gene Symbol: Elf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012181.2 | NP_001012181.1 | 7 | 2135 | ||
NM_001033909.1 | NP_001029081.1 | 9 | 1815 | |||
XM_006232325.2 | XP_006232387.1 | 10 | 2017 | |||
XM_006232327.2 | XP_006232389.1 | 7 | 4203 | |||
XM_006232328.1 | XP_006232390.1 | 8 | 1529 | |||
XM_006232329.1 | XP_006232391.1 | 8 | 1493 | |||
XM_008760992.1 | XP_008759214.1 | 9 | 2400 | |||
GenBank mRNA | BC083585.1 | - | 10 | 1967 | 7131145 | |
BC083598.1 | - | 7 | 1487 | 7132941 |
