siRNA Details
Gene Symbol: Postn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108550.1 | NP_001102020.1 | 2 | 156 | ||
XM_006232335.2 | XP_006232397.1 | 2 | 183 | |||
XM_006232336.2 | XP_006232398.1 | 2 | 182 | |||
XM_006232337.2 | XP_006232399.1 | 2 | 181 | |||
GenBank mRNA | KM117173.1 | - | 2 | 138 |
