siRNA Details
Gene Symbol: Veph1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014171.1 | NP_001014193.1 | 9 | 1834 | ||
XM_008761050.1 | XP_008759272.1 | 8 | 1797 | |||
XM_008761051.1 | XP_008759273.1 | 9 | 1834 | |||
GenBank mRNA | BC087055.1 | - | 9 | 1834 | 7123921 |
