siRNA Details
Gene Symbol: Veph1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014171.1 | NP_001014193.1 | 3 | 489 | ||
XM_008761050.1 | XP_008759272.1 | 2 | 452 | |||
XM_008761051.1 | XP_008759273.1 | 3 | 489 | |||
XM_008761052.1 | XP_008759274.1 | 2 | 452 | |||
XM_008761053.1 | XP_008759275.1 | 2 | 452 | |||
GenBank mRNA | BC087055.1 | - | 3 | 489 | 7123921 |
