siRNA Details
Gene Symbol: Lrba | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108555.1 | NP_001102025.1 | 17, 18 | 2162 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Lrba | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108555.1 | NP_001102025.1 | 17, 18 | 2162 |