siRNA Details
Gene Symbol: Dennd4b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008761246.1 | XP_008759468.1 | 2 | 2366 | ||
XM_008761247.1 | XP_008759469.1 | 2 | 2366 | |||
XM_008761248.1 | XP_008759470.1 | 3 | 1611 | |||
XM_008761249.1 | XP_008759471.1 | 3 | 1053 | |||
XM_342287.8 | XP_342288.6 | 3 | 1611 |
